missing translation for 'onlineSavingsMsg'
Learn More

SPEX CertiPrep™ Pulverizer and Cell Lyser 2010 Geno/Grinder

Product Code. 12605297 Shop All SPEX Certiprep Products
Click to view available options
Dimensions (L x W x H):
57.2 x 35.6 x 71.1 cm
Unit Size:
Each
This item is not returnable. View return policy
Récupel-bijdrage uitgesloten

Product Code. 12605297

Brand: SPEX CertiPrep™ 2010230

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Récupel-bijdrage uitgesloten

Automated high-throughput plant and animal tissue homogenizer and cell lyser. SPEX CertiPrep™ Pulverizer and Cell Lyser 2010 Geno/Grinder accommodates a full range of sample vials or up to six deep-well titer plates.

  • Automated mechanical disruption through bead beating, ideal for high-throughput cell lysis applications.
  • Applications:
    • Isolation of nucleic acids (DNA/RNA), proteins, enzymes, bacteria and yeasts.
    • Extraction of pesticides and other low level residues from plant and animal samples.
    • Used in modified QuEChERS, SweET and other extraction methods.
  • Specifically designed for rapid cell disruption, lysis and tissue homogenization.
  • Designed for vigorous up-and-down shaking for quick and effective sample preparation (typically within 2 minutes) from deep-well titer plates, vial sets, and centrifuge tubes.
  • Programmable touch screen control panel for run time, rate, cycles and pause time.
  • Up to 500 protocols can be saved for recall.
  • Adjustable clamp with a release button that allows users to secure vials or titer plates of various sizes.
  • Safety interlock and pneumatic cylinder stabilize the lid for safe operation.
  • Horizontal clamp loading prevents sample spillage and eliminates cross-contamination.
  • Simple to set up and load - just load samples into the clamp, set speed and time and press the start button for quick and consistent sample preparation.
  • Optional Kryo-Tech™ accessories enable cryogenic grinding and preserve temperature sensitive samples for RNA and protein extractions.

Specifications

Certifications/Compliance CE Approved
Dimensions (L x W x H) 57.2 x 35.6 x 71.1 cm
Grinding Time Max: 20 min.
Hertz 50 Hz
Horsepower 1/3 hp motor
Includes Universal clamp and nesting tray for use with multiwell plates
Plug Type 2-prong grounded plug
Voltage 230 V
Weight (Metric) 45.5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.