Learn More
Wheaton™ PTFE/Silicone Septa
Brand:  Wheaton™ W240583A
136.53 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11724239
  • 140.00€ / Pack of 100
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Specifications
13 mm
Autoclavable
Septa
PTFE, Silicone
100 Pk.
SDS
Documents Certificates
Product Title
Wheaton™ PTFE/Silicone Septa > Outer Diameter: 13mm