missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Plain Stirrer Shaft
Fisherbrand™ PTFE Plain Stirrer Shafts have a PTFE encapulated stainless steel core with exposed end. They are a fully molded construction featuring a PTFE roto molded to a fully PTFE encapsulated steel reinforced drive shaft.
Brand: Fisherbrand™ 005.1016750EX
Description
- PTFE with Steel Core
- 50mm Exposed End
- Chemically Inert
- Easy to clean
- No cross contamination
- Non scratch
- Use up to 280° C
Specifications
PTFE Plain Stirrer Shaft | |
750 x 16 mm | |
1 |
10 mm | |
750 mm | |
16mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction