missing translation for 'onlineSavingsMsg'
Learn More

PSB 0788, Tocris Bioscience™

Product Code. 18014232
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective A2B antagonist

Potent and selective adenosine A2B receptor antagonist (IC50 = 3.64 nM at human A2B receptors). Displays > 100-fold selectivity over hA1, hA2A and hA3 receptors.

Chemical Identifiers

CAS 1027513-54-7
Molecular Formula C25H27ClN6O4S
Molecular Weight (g/mol) 543.039
InChI Key JQZJACVYMPKVDS-UHFFFAOYSA-N
Synonym d05xgf, 8-4-4-4-chlorobenzyl piperazine-1-sulfonyl phenyl-1-propylxanthine, 8-4-4-4-chlorobenzyl piperazide-1-sulfonyl phenyl-1-propylxanthine, 8-4-4-4-chlorobenzyl piperazin-1-ylsulfonyl phenyl-1-propyl-1h-purine-2,6 3h,7h-dione, 8-4-4-4-chlorophenyl methyl piperazine-1-sulfonyl phenyl-1-propyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione
PubChem CID 44186581
IUPAC Name 8-[4-[4-[(4-chlorophenyl)methyl]piperazin-1-yl]sulfonylphenyl]-1-propyl-3,7-dihydropurine-2,6-dione
SMILES CCCN1C(=O)C2=C(NC1=O)N=C(N2)C3=CC=C(C=C3)S(=O)(=O)N4CCN(CC4)CC5=CC=C(C=C5)Cl

Specifications

Quantity 10 mg
Formula Weight 543.04
Percent Purity >98%
Chemical Name or Material PSB 0788
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'