Learn More
Mutexil™ Protective Overshoe Coverings
Click to view available options
Product Type:
Overshoe
Protective CPE Visitor Overshoes
Sticky Overshoe
Color:
Blue
Transparent
White
Size:
2X-Large
Universal
X-Large
Unit Size:
25 pairs
Pack of 1000
Pack of 2000
Pack of 300
Description
- Various sizes and colors
- Needs-based styles
- Per new standards EN ISO 20345 and EN ISO 20347
Specifications
Specifications
| Color | White |
| For Use With (Application) | For sanitary, food, industrial, catering and services |
| Material | Polypropylene |
| Size | Universal |
| Style | CPE |
| Product Type | Protective CPE Visitor Overshoes |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion