missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ProSpecT™ Entamoeba histolytica Microplate Assay
Rapidly detect Entamoeba histolytica specific antigen from human fecal samples.
Brand: Thermo Scientific™ R2456096
1069.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Rapidly detect Entamoeba histolytica specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Entamoeba histolytica Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Entamoeba histolytica. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
ProSpecT Entamoeba histolytica is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Entamoeba histolytica provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
Yes | |
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card |
ProSpecT Entamoeba histolytica Microplate Assay | |
E. histolytica Antigen | |
96 Tests/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction