Learn More
Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay
Rapidly detect Crytosporidium specific antigen from human fecal samples.
Brand:  Thermo Scientific™ R2454096
1117.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11404059
  • 1274.00€ / 96 tests
Explore more special offers
Description
Description
Rapidly detect Crytosporidium specific antigens (CSA) in fresh or preserved fecal specimens using Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Crytosporidium. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Cryptosporidium is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures, and common reagents, maximizing operational efficiency, productivity and throughput.
ProSpecT Cryptosporidium provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2°C to 8°C
Cryptosporidium Antigen
96 Wells/Kit
ProSpecT Cryptosporidium Microplate Assay
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6212063 Certificate TypeCertificate of Analysis Date25/03/2025 Catalog NumberR2454096
Lot Number6212062 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog NumberR2454096
Lot Number6184248 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberR2454096
Lot Number6162865 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog NumberR2454096
Lot Number6152219 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog NumberR2454096
Product Title
Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay > Prspct Crypto Micrplat 96pk