missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ proFood™ 79-340 Series Green Nitrile Gloves

Reusable gloves suitable for contact with all fatty foodstuffs.

Brand:  Ansell™ 79-340_9.5-10

 View more versions of this product

Product Code. 13407556

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Good mechanical and chemical properties
  • Contains no natural rubber
  • Safe for both food and workers hands
  • Flexibility, even in cold condition

Applications:

  • Poultry processing
  • Beef and pig processing
  • Fish processing
  • Janitorial use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nitrile
Gloves
79-340
Green
Cotton Flocking
Mechanical and chemical
Nitrile
10
0.3 mm
Straight
12 Pairs
79-340
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.