missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ ProfiLine™ pH 3210 Portable pH Meter
Brand: WTW™ 2AA214
This item is not returnable.
View return policy
Description

Specifications
Carry case with pH electrode SenTix™ 51, Buffer solution STP 4 and STP 7, stand, beaker, short instruction manual, CD-ROM and batteries | |
±0.3, ±1 | |
For challenging measurements in the field and plant, e.g. water/wastewater analysis, chemical/pharmaceutical industry, etc | |
±0.005 | |
±0.3, ±1mV | |
LCD | |
CE | |
80 W x 180 H mm | |
3 Years | |
4 x 1.5V AA or 4 x 1.2V NiMH rechargeable | |
-5°C to +105°C | |
Portable pH meter | |
80 W mm | |
180 mm |
±0.1°C | |
±0.1, ±0.01, ±0.005 | |
-2 to 19.999 | |
-1200 to 1200, -2500 to 2500 | |
1, 2, 3, 4, 5 Calibration WTW Technical Buffer, DIN/NIST buffers plus 20 additional buffer set | |
Manual 200 | |
IP-66/67 | |
300 g | |
pH 3210 SET 4 | |
All values ±1 digit | |
AA, NiMH | |
1000 hr. | |
80 mm | |
55 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction