missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ ProfiLine™ pH 3110 Portable Meter
Brand: WTW™ 2AA111
This item is not returnable.
View return policy
Description
Specifications
Carry case with pH electrode SenTix™ 21, Buffer solution STP 4 and STP 7, stand, beaker, short instruction manual, CD-ROM and batteries | |
±0.3, ±1 | |
Ideal for all seeking an easy, robust and waterproof instrument for portable pH measuring | |
±0.005 | |
±0.3, ±1mV | |
LCD, 7 Segment | |
IP-67 | |
0.6 kg | |
pH 3110 SET 1 | |
All values ±1 digit | |
AA, NiMH | |
2500 hr. | |
80 mm | |
55 mm |
±0.1°C | |
±0.1, ±0.01, ±0.005 | |
-2 to 19.999 | |
-1200 to 1200, -2000 to 2000 | |
1, 2 or 3 Calibration WTW Technical Buffer, DIN/NIST buffers | |
CE | |
80 W x 180 H mm | |
3 Years | |
4 x 1.5V AA or 4 x 1.2V NiMH rechargeable | |
-5°C to +105°C | |
Portable pH meter | |
80 W mm | |
180 mm |