Learn More
WTW™ ProfiLine™ Oxi 3210 Portable Dissolved Oxygen Meter
A versatile, portable, comfortable DO-meter, the ProfiLine Oxygen Meter Oxi 3210 has a backlit LCD graphic display for mobile measurement. For all portable field measurement applications.
Brand:  WTW™ 2BA201
Product Code. 10698384
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Specifications
Meter with carry case including galvanic DO probe CellOx™ 325, cleaning solution, electrolyte, 3 exchange membrane heads, stand, beaker, instruction manual, CD-ROM and batteries
500 to 1100mbar
-5°C to +105°C
±0.1°C
200 Manual points
IP-67
4 x 1.5V AA or 4 x 1.2V NiMH batteries
0 to 90mg/L
0.01, 0.1mg/L
1%
NiMH rechargeable
17.7 cm
5.7 cm
For all portable field measurement applications
1
0.1
LCD
cETLus, CE, ISO 9001:2008
3 Years
Galvanic DO probe, CellOx 325
0 to 70ppt
0 to 600%
Up to 1000hr.
Portable Dissolved Oxygen Meter
7.9 cm
±0.5% of value