Learn More
Liebherr™ ProfiLine Forced-Air Freezer
Use these freezers for general laboratory storage of temperature-sensitive samples. Liebherr™ ProfiLine Forced-Air Freezers feature chrome-nickel-steel interior containers and plastics coated grid shelves.
Brand:  Liebherr™ GGPv 1470
 View more versions of this product
Product Code. 12180162
Cet article n'est plus commercialisé et n'est donc pas disponible. Consultez le produit pour voir les alternatives possibles ou contactez notre support technique au 056 260 260.
View alternative products »
Explore more special offers
Description
Description
  • Gastronorm ProfiLine recirculated air freezer
  • Steel SwingLine design door
  • Fitted lock
  • Electronic control system
  • External digital temperature display
Specifications
Specifications
1427 L
White
215 H x 143 W x 83 D cm
155 cm
220/240 V
R290
-10°C to -26°C
GGPV1470X
65 cm
124 cm
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products