missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ ProfiLine™ Cond 3110 Portable Conductivity Meter
Brand: WTW™ 2CA304
This item is not returnable.
View return policy
Description
Specifications
Meter with carry case including 2-electrode conductivity cell LR 325/01 including flow-through vessel, stand, beaker, short instruction manual, CD-ROM and batteries, driver software for USB, cable | |
For professional field measurements requiring documentation and data transfer | |
1, 2, 3-point WTW Technical and NIST Buffer | |
0.001, 0.01, 0.1 mS | |
0 to 70 ppt | |
±0.2% Full-scale | |
±0.1°C | |
CE | |
0.6 kg | |
Cond 3110 | |
-5°C to +105°C | |
17.7 cm | |
5.7 cm |
±0.5% Full-scale ±1 LSD | |
Fixed: 0.475cm–1/1cm–1, Calibratable: 0.450 to 0.500cm–1/0.585 to 715cm–1/0.800 to 1.200cm–1 | |
0 μS to 1000 mS | |
Automatic | |
0.1 ppt | |
0.1 | |
LCD | |
IP67 | |
3 Years | |
4 x 1.5V AA oder 4 x 1.2V NiMH-rechargeable battery | |
Portable Conductivity Meter | |
7.9 cm | |
NiMH |