missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Profaviton BV-06 Chemical Protection Gloves

Product Code. 17260511
Click to view available options
Size:
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical protection gloves without stockinette constructed of butyl rubber, with a seamless bromobutyl coating. Viton™ outer layer is resistant to aliphatic and aromatic hydrocarbons, halogenated hydrocarbons, organic/inorganic acids, and saturated saline solutions.

  • Butyl rubber chemical protection gloves without stockinette
  • Seamless, bromobutyl coating is 0.40 mm thick
  • Viton™ outer layer is resistant to a wide variety of chemicals
  • Enhanced grip in damp or wet areas
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material Bromobutyl, Viton
Color Black
Size 9
EN388 Standard 2120A
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AFKLMN
For Use With (Application) Chemical Handling
Gender Unisex
Glove Cuff Style Gauntlet
Series Profaviton BV-06
Length (Metric) 35 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'