Learn More
Invitrogen™ eBioscience™ ProcartaPlex Rat IL-10 Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand: Invitrogen™ EPX01A-36049-901
Includes:
- concentrated Magnetic Beads coated with monoclonal antibody to rat IL-10
- rat IL-10 Standard (lyophilized)
- Biotin-Conjugate (50x) anti-rat IL-10 monoclonal antibody
Descripción
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Especificaciones
96 Tests | |
Multiplex Immunoassay | |
Serum, Plasma: 25 μL; CCS: 50 μL |
Rat | |
Cell Culture Supernatant, Plasma, Serum | |
Interleukin-10, TGIF, MCSF III, CSIF, B TCGF |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido