missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Prolex™ - Blue E. coli 0157 Latex Test Reagent Kits
Just one colony is sufficient for the test, enabling even limited growth to be detected
51.00€
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Clia Complexity | Non-waived |
Detectable Analytes | E. coli serogroup O157 |
Sensitivity | 1 |
Type | Blue E. coli O157 Latex Test Reagent Kit |
Product Code | Brand | Quantity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Includes | Price | Quantity & Availability | ||||
11969044
|
Pro-Lab Diagnostics™
PL.070B |
50 Tests | E. coli O157 Latex reagent 3.1mL, negative control Latex reagent 1.5mL, positive control 1.5mL, Agglutination cards 25, mixing sticks 75 |
51.00€
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Developed to remove the possibility of cross-reactions with E. hermanii and other sorbitol negative Escherichia species
- Latex agglutination
- Test results in 2 minutes
- CLIA Complexity: Non-waived
- Sensitivity of 100%
Specifications
2°C to 8°C | |
E. coli serogroup O157 | |
Blue E. coli O157 Latex Test Reagent Kit |
Non-waived | |
1 | |
2 min. |
Safety and Handling
missing translation for 'shelfLife' : 2 Years
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title