Learn More
Zeiss™ Primostar 3
Robust and compact upright microscope for digital teaching and routine lab work. Full Kohler stand for transmitted brightfield microscopy and with a phototube for the addition of a camera.
Brand:  Zeiss™ 415501-­0041-­000
2074.81 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 17244443
  • 2690.00€ / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Description
  • The Primostar 3 is a robust yet compact upright microscope for digital teaching and routine lab work. The Primostar 3 is reliable and robust due to a study metal frame, whilst being light weight. The Primostar 3 has also been designed for maximum ease of use.
  • Fixed-Köhler - ideal in the Educational setting.
  • The Fixed-Köhler versions of Primostar 3 come pre-adjusted with a field of view of 20 mm. Selected objective lenses and eyepieces are already in place meaning no time is wasted in the set up. Simply plug in your microscope and start your lesson.
  • Full-Köhler - ideal in teaching and routine labs
  • Full-Köhler versions of Primostar 3 with field of view 22 mm give freedom to teach while using Köhler illumination. In pre-defined packages, a selection of objective lenses and eyepieces are already in place. Benefit from a relaxed posture for day-long work with enlarged stage drive. If you have more specimens in place, you can use the double slider holder.
Specifications
Specifications
Phototube and a Full Kohler Stand