Learn More
Invitrogen™ PRIMER COP 200 NMOL
Custom Primer
Brand:  Invitrogen™ 10913044
24.06 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10615933
  • 32.08€ / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
COP Primer, 200 nmol
COP Primer
200 nmol