Learn More
Invitrogen™ PRIMER 5'PHO 50NMOLE PLATE
Custom Primer
Brand:  Invitrogen™ 10625119
8.94 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10286542
  • 11.92€ / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' PHO Primer Plate, 50 nmol
Room Temperature
50 nmol
5' PHO Primer Plate
5' Labeled