Learn More
Invitrogen™ PRIMER 5' BIOTIN 1 UMOL
Custom Primer
Brand: Invitrogen™ 10626026
231.75 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
5' Biotin Primer, 1μmol | |
5' Biotin Primer | |
5' Labeled | |
1 μmol |