Learn More
Invitrogen™ PRIMER, 5'ALEXA 514 200 NM
Custom Primer
Brand: Invitrogen™ 100007344
163.15 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
5' Alexa 514 Primer, 200 nmol | |
5' Alexa 514 Primer | |
5' Labeled | |
200 nmol |