Learn More
Invitrogen™ PRIMER, 5'ALEXA 488 50 NMOL
Custom Primer
Brand:  Invitrogen™ 10660000
144.00 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10276152
  • 192.00€ / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' Alexa 488 Primer, 50 nmol
5' Alexa 488 Primer
5' Labeled
50 nmol