Learn More
Invitrogen™ PRIMER,3'PHO 200 NMOL
Custom Primer
Brand: Invitrogen™ 12499034
78.67 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
3' PHO Primer, 200 nmol | |
3' PHO Primer | |
3' Labeled | |
200 nmol |