Learn More
Tocris Bioscience™ PRIMA-1MET
Restores mutant p53 activity
Brand:  Tocris Bioscience™ 3710/10
Product Code. 18035722
  • 189.00€ / 10mg
Explore more special offers
Description
Description
PRIMA-1MET is a methylated derivative of PRIMA-1 (Cat. No. 1862). Restores mutant activity p53 activity. Acts synergistically with chemotherapeutic agents to inhibit tumor cell proliferation.
Specifications
Specifications
>98%
10 mg
199.25
APR-246
PRIMA-1MET
PRIMA-1MET