Learn More
Tocris Bioscience™ PRIMA-1MET
Restores mutant p53 activity
Brand: Tocris Bioscience™ 3710/10
This item is not returnable.
View return policy
Description
PRIMA-1MET is a methylated derivative of PRIMA-1 (Cat. No. 1862). Restores mutant activity p53 activity. Acts synergistically with chemotherapeutic agents to inhibit tumor cell proliferation.Specifications
>98% | |
10 mg | |
199.25 |
APR-246 | |
PRIMA-1MET | |
PRIMA-1MET |