missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Precision Hydrometers for Specific Gravity

Product Code. 11766923
Encompass_Preferred
Click to view available options
Specific Gravity Range:
0.590 to 0.660
1.200 to 1.420
1.400 to 1.620
1.600 to 1.670
1.660 to 1.730
Increments:
0.0005 sp.gr.
0.002 sp.gr.
Length (Metric):
300 mm
325 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11766923

Brand: Fisherbrand™ 2571DBQ

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

High-precision instruments for determination of density in fluids. Fisherbrand™ Precision Hydrometers for Specific Gravity do not include a thermometer in the body and feature a reference temperature of 60/60°F.

High-precision instruments for determination of fluid density available with a DAkkS or works calibration certificate.
  • Reference temperature: 60/60°F
  • Without thermometer – control thermometer sold separately
  • Single packed in transparent square plastic sleeve

Specifications

Type Hydrometer without Thermometer
Accuracy ±0.0005 sp/gr
Increments 0.0005 sp.gr.
Length (Metric) 325 mm
For Use With (Equipment) Control thermometer
Includes Plastic sleeve
Material Glass
For Use With (Application) Measuring specific gravity
Specific Gravity Range 1.660 to 1.730
Temperature of Standardization 60°F
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.