Learn More
Buerkle™ PP Lid for Buerkle™ Storage/Stacking Container
Buerkle™ PP Lid for Buerkle™ Storage/Stacking Container is the perfect solution to cover the Storage/Stacking Containers.
22.25€ - 36.35€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10268920
|
Buerkle™
3414-0001 |
400 x 300 mm Storage/Stacking Containers |
22.25€
Each |
In Stock
Log in to see stock. |
|||||
10441561
|
Buerkle™
3414-0002 |
600 x 400 mm Storage/Stacking Containers |
36.35€
Each |
In Stock
Log in to see stock. |
|||||
Description
- PP
- Lid for storage containers
Specifications
Lid | |
Polypropylene | |
Lid |