Learn More
3M™ Powerflow™ Full Facepiece 7900
3M™ Powerflow™ Full Facepiece 7900 is a replacement full-face mask for 3M™ Powerflow™ 7900PF Powered Air Respirator.
Brand:  3M™ 7000103984
Product Code. 15507469
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Replacement full face mask for 3M™ Powerflow™ 7900PF Powered Air Respirator
  • Speech diaphragm to improve communication
  • For use with 3M™ Powered Air Respirator 7900PF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN 136:1998 (Class 2), CE Approved
Silicone, Polycarbonate
Face-mounted
Black
3M™ Powerflow™ 7900PF Powered Air Respirator