Learn More
Knick Elektronische™ Portamess™ 911 Conductivity Meter
Basic unit for mobile conductivity measurement. Includes easy-to-read LC display with antireflection natural glass.
Brand:  Knick Elektronische™ 904 Cond
Product Code. 12047232
  • 2230.00€ / Each
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Description
  • Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
  • 15mm character height LC display with antireflection natural glass is scratchproof and resistant to chemicals
  • Clear icons for operator guidance
  • Integrated quiver protects the sensor and is replaceable. Transparent and easy to remove
  • Blue ergonomic enclosure is resistant to impact and chemicals with watertight, diecast chassis (IP 66 protection)
  • Cover protects against dirt and damage.
  • Highly flexible lamellar joint can be folded back completely.
  • Fold-out hook for hanging or standing up. Keeps both hands free for handling the sensor.
  • Gold-plated, water-tight robust sockets
  • Practical, adjustable carrying strap
Specifications
Specifications
Mobile conductivity measurement
911
160 mm
LC
Smooth membrane keypad
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products