Learn More
Thermo Scientific™ POROS™ 50 HS Strong Cation Exchange Resin
Thermo Scientific™ POROS™ 50 micron bulk resins are available for low pressure applications, including large scale downstream bioprocessing.
646.00€ - 3660.00€
Specifications
Particle Size | 50 μm |
---|---|
Pore Size | 500 to 10,000 Å |
Stationary Phase | HS |
Shipping Condition | Room Temperature |
Product Line | POROS™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019650
|
Thermo Scientific™
1335906 |
50 mL |
646.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019888
|
Thermo Scientific™
1335911 |
250 mL |
1758.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30067385
|
Thermo Scientific™
1335907 |
1 L |
3660.00€
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POROS 50 micron bulk resins are available for low pressure applications, including large scale downstream bioprocessing.
POROS High Performance Chromatography™ Media and Pre-Packed Columns
A high performance chromatography resin for analytical to process scale separations.
- Higher productivity: High throughput and high dynamic capacity associated with High Performance Chromatography™
- Chemical stability: Allows aggressive cleaning and sanitization
- Enhanced biomolecule access: Provided via large pores, ranging between 500-10,000 Å
- Polystyrenedivinylbenzene particles: Yield a robust, easily packable matrix
- Develop better separations methods in a shorter time frame: The speed of High Performance Chromatography technology reduces weeks of experimentation to only a few hours of work, so you have plenty of time to explore all the variables of your separation
Reduce time-consuming sample prep
You can replace many sample preparation steps with high speed High Performance Chromatography technology. For instance, dialysis of large volumes of material can be replaced by high flow rate processing of the dilute sample. Elution in a small volume of new buffer accomplishes both buffer exchange and concentration.
Create novel assays for more efficient analysis
High Performance Chromatography technology is not limited to standard modes of separation. Both enzymes and affinity ligands can be immobilized on POROS media. By combining rapid on-column protein digestions with rapid on-column immunoassays and chromatographic separations, you can create entirely new assays with unlimited potential.
High capacity, high resolution, high speed
In contrast to conventional chromatography media, POROS High Performance Chromatography™ resins particles, are engineered to have two discreet classes of pores. Large 'throughpores' allow convection flow to occur through the particles themselves, quickly carrying sample molecules to short 'diffusive' pores inside. By reducing the distance over which diffusion needs to occur, the time required for sample molecules to interact with interior binding sites is also reduced. Diffusion is no longer limiting and flow rates can be dramatically increased - without compromising resolution or capacity. Separations can be achieved at 1000 to 5000 cm/hr compared to 50 to 360 cm/hr for conventional media.
Specifications
50 μm | |
HS | |
POROS™ |
500 to 10,000 Å | |
Room Temperature | |
Strong Cation Exchange |
Safety and Handling
Recommended Storage : Room Temperature