Learn More
Rogo-Sampaic Porcelain Buchner Funnels
Excellent for separating liquids from solids in the laboratory. Porcelain Buchner Funnels provide a quick and easy pressure assisted filtration method.
Brand:  Rogo-Sampaic PNF011
 View more versions of this product
Product Code. 11717335
  • 101.00€ / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • White porcelain
  • Available for a range of volumes and filter diameters
  • Applications include collecting recrystalized compounds
  • Excellent thermal and chemical resistance
Specifications
Specifications
265 mL
90mm
71mm
Buchner Funnel
Porcelain
90mm
1Each