missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-7™ Polymer for Protein Quality Analysis

Product Code. p-7138023 Shop All Applied Biosystems Products
Click to view available options
Quantity:
384 Samples
960 Samples
Unit Size:
Each
Pack of 384
Pack of 960
This item is not returnable. View return policy

Product Code. 16326512

Brand: Applied Biosystems™ A30936

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Includes

Radio frequency identification (RFID) label.

This is a pouch of POP-7 polymer for protein quality analysis on 3500/3500xL genetic analyzers. The POP-7 separation matrix is validated for protein quality analysis and is offered in easy-to-use pouch packages and 960 sample (maximum) or 384 sample (maximum) size formats.

  • Easy to use: Pre-formulated and in a single-use pouch, which not only saves time but ensures reliability and consistency
  • Robust formulation: Formulated for biomolecular analysis applications
  • Replenishable: Allows capillaries to be used multiple times
  • Includes radio frequency identification (RFID) label: Easily track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2–8°C.
Description POP-7™ Polymer For 3500xL Genetic Analyzer
For Use With (Equipment) 3500xL Genetic Analyzer for Protein Quality Analysis
For Use With (Application) Glycan Analysis, Protein Quality Analysis
No. of Samples 384
Product Line POP-7
Product Type Polymer
Quantity 384 Samples
Shipping Condition Wet Ice

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.