Learn More
Tocris Bioscience™ Pomalidomide 4'-PEG2-azide
Cereblon ligand with PEG linker and terminal azide for onward chemistry
Brand:  Tocris Bioscience™ 7307/25
204.74 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18717216
  • 260.00€ / 25mg
Explore more special offers
Description
Description
Functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG2 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
4-[[2-[2-(2-Azidoethoxy)ethoxy]ethyl]amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione
25 mg
C19H22N6O6
2271036-45-2
Degrader Building Blocks
0.95