Learn More
Kautex™ PVC Wide Neck Square Container, Transparent, Without Closure
Fill and clean these wide neck PVC containers with ease due to the roomy neck configuration. Supplied without closure.
Brand:  Kautex™ 2000073352
 View more versions of this product
Product Code. 17838127
  • 197.00€ / Pack of 44
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Explore more special offers
Description
Description
  • Member of Series 310 container family
  • Closure available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2000 mL
69 mm
PVC
44/Cs.
Square
Wide Neck Container
Transparent
Packaging, Dispensing, Sending Samples, Storage, Mailing
44
No
187 mm
Closure, Foam Insert