missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ PVC Wide Mouth Square Containers

Kautex–PVC Wide Mouth Square Containers–Manufactured with crystal-clear PVC for optimum visibility and excellent product presentation

Brand:  Kautex™ 2000070595

Product Code. 17818127

  • 610.00€ / Pack of 972

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Easy to fill and to clean
  • Square shape optimizes space utillization
Application:
  • packaging
  • dispensing
  • sending samples
  • storage
  • mailing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mL
38.5 x 38.5 x 59 mm
Packaging, Dispensing, Sending Samples, Storage, Mailing
972
No
Wide Neck Container
Transparent
23 mm
PVC
972/Cs.
Square
Closure, Foam Insert
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions