missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Polyurethane Sani-Sponge with Dey-Engley Neutralizing Broth
Synthetic polyurethane sponge for sampling surfaces, with buffer to neutralize disinfectant.
258.00€ - 301.00€
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17976679
|
LABPLAS
KSS-67810-DE |
None |
258.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17956509
|
LABPLAS
KSS-67815-DE |
Gloves |
301.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Polyurethane Sani-Sponge
- Saturated with Dey-Engley neutralizing broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit |