missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polystyrene Petri Dishes

Manufactured from highly transparent polystyrene for its heat resistant properties to enable use with hot agar. Greiner BioOne™ Polystyrene Petri Dishes are available in a wide variety of formats and sizes to accommodate multiple research needs.

Brand:  Greiner Bio-One 639102

 View more versions of this product

Product Code. 10340111

  • 108.00€ / Pack of 120
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Heat resistant up to 60°C
  • Available in vented and non-vented
  • Choose from standard round design, square-profile dish, special models and contact dishes/OneWell Plate™ models
  • Easy stacking
  • Crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Round
120 Pack
20 mm
Polystyrene
Non-sterile
Yes
145 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.