Learn More
Greiner Bio-One Polystyrene Conical Bottom Analyzer Cups
Greiner Bio-One Cell 1.5mL Polystyrene Conical Bottom Analyzer Cups suitable for use with analytical systems, such as Technicon, Gemsaec, Centrifichem, Hitachi, TOA and Coulter analyzers.
Brand:  Greiner Bio-One 621171
 View more versions of this product
Product Code. 10660742
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Material: Polystyrene
  • Conical bottom
  • With support skirt
  • For single use only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Technicon Analyzer
Non-sterile
Polystyrene
4 mL
Conical
Clear
Analyzer Cup