Learn More
Greiner Bio-One Polystyrene 96-well Half Area Microplates
Reduces sample volume up to 50%. Greiner Bio-One™ Polystyrene 96-Well Half Area Microplates provide the ease of manual pipetting.
299.00€ - 682.00€
Specifications
Certifications/Compliance | DIN ISO 9001 |
---|---|
Certified | Yes |
Format | Half-Area |
Array | 8 x 12 |
Binding Type | Medium Binding |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10791624
|
Greiner Bio-One
675076 |
Black |
299.00€
Pack of 40 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
10115853
|
Greiner Bio-One
675096 |
Clear |
682.00€
Pack of 40 |
Estimated Shipment: 05-03-2025
Log in to see stock. |
|||||
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Traceable to production through a defined LOT number system
- μClear™ bottom
Specifications
DIN ISO 9001 | |
Half-Area | |
Medium Binding | |
96 | |
Yes |
Yes | |
8 x 12 | |
Polystyrene | |
Yes |