Learn More
Greiner Bio-One Polystyrene 96-well Half Area Microplates
Reduces sample volume up to 50%. Greiner Bio-One™ Polystyrene 96-Well Half Area Microplates provide the ease of manual pipetting.
299.00€ - 682.00€
Specifications
Certifications/Compliance DIN ISO 9001
Certified Yes
Format Half-Area
Array 8 x 12
Binding Type Medium Binding
View More Specs
Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10791624
View Documents
Greiner Bio-One
675076
Black
299.00€
Pack of 40
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
10115853
View Documents
Greiner Bio-One
675096
Clear
682.00€
Pack of 40
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Traceable to production through a defined LOT number system
  • μClear™ bottom
Specifications
Specifications
DIN ISO 9001
Half-Area
Medium Binding
96
Yes
Yes
8 x 12
Polystyrene
Yes