Learn More
BRAND™ Polypropylene Powder Funnels
Constructed of durable, lightweight polypropylene. Brand™ Polypropylene Powder Funnels are an excellent option for the transfer of samples without flow rate restrictions.
5.04€ - 83.47€
Product Code | Brand | Diameter (Metric)Top | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Height (Metric) | Price | Quantity & Availability | |||||
11633139
|
BRAND™
|
80 mm | 26 mm (Stem) | N/A | ||||||
17400134
|
BRAND™
|
80 mm | 26 mm (Stem) |
49.19€
Pack of 10 |
In Stock
Log in to see stock. ![]() |
|||||
11653139
|
BRAND™
|
120 mm | 28 mm (Stem) | N/A | ||||||
17420134
|
BRAND™
|
120 mm | 28 mm (Stem) |
57.95€
Pack of 10 |
Estimated Shipment: 04-04-2025
Log in to see stock. |
|||||
17430134
|
BRAND™
|
150 mm | 42 mm stem |
37.23€
Pack of 5 |
In Stock
Log in to see stock. ![]() |
|||||
10748431
|
BRAND™
|
150 mm | 42 mm stem | N/A | ||||||
10777591
|
BRAND™
|
180 mm | 48 mm (Stem) |
13.05€
Each |
In Stock
Log in to see stock. ![]() |
|||||
11623139
|
BRAND™
148020 |
65 mm | 26 mm (Stem) | N/A | ||||||
17490124
|
BRAND™
148020 |
65 mm | 26 mm (Stem) |
25.65€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- With short, wide stem
- Provides excellent resistance against thermal shock
Specifications
Polypropylene | |
Autoclavable | |
Translucent |
Product Title
BRAND™ Polypropylene Powder Funnels