Learn More
Greiner Bio-One Polypropylene Microcentrifuge Tubes
For use in safe and long term storage. Greiner Bio-One Polypropylene Microcentrifuge Tubes offer a leak-free performance and minimal non-specific binding.
85.65€ - 1025.00€
Product Code | Brand | Includes | Closure Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Closure Color | Price | Quantity & Availability | ||||
11857822
|
Greiner Bio-One
716201 |
N/A | Natural | N/A | |||||
10111852
|
Greiner Bio-One
717261 |
Support skirt | White |
1025.00€
Pack of 3000 |
Estimated Shipment: 03-03-2025
Log in to see stock. |
||||
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- chemical and temperature resistance
- clarity coupled to a wide thermal range
- May be centrifuged to 20,000g