accessibility menu, dialog, popup
Test
chevron-down
Select Language
English
French
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Chemical Hazard Signs
Brady™ Polypropylene Laser Radiation Warning Sign
Print
Brady™ Polypropylene Laser Radiation Warning Sign
10.95€ - 25.25€
Specifications
Shape
Triangular
View More Specs
Products
2
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
2
Product Code
Brand
Width (Metric)
Price
Quantity & Availability
Product Code
Brand
Width (Metric)
Price
Quantity & Availability
11816371
View Documents
missing translation for 'viewProductCertificates'
Brady™
252218
200 mm
10.95€
Each
In Stock
Log in
to see stock.
Add to basket
11754844
View Documents
missing translation for 'viewProductCertificates'
Brady™
252219
315 mm
25.25€
Each
Estimated Shipment: 18-12-2024
Log in
to see stock.
Add to basket
Description
Description
Specifications
Specifications
Shape
Triangular
Videos
SDS
Documents
Documents
Product Certifications
Back to Top