missing translation for 'onlineSavingsMsg'
Learn More

Haleco™ Polypropylene Chemical Absorbent Sheet

An excellent absorption solution for most aggressive chemicals including acids, bases, alcohol, ketones and more.

Brand:  Haleco™ CPA2151H

Product Code. 12636316

  • 243.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Manufactured from 100% polypropylene
  • Honeycomb structure for greater mechanical resistance
  • Double thickness
  • Supplied in a dispensing box for more practical storage and handling
  • Incinerable: ash index less than 0.05%
  • 100 sheets per box
  • 50 x 40cm in size
Specifications

Specifications

Yellow
Polypropylene
Absorption of hazardous liquids and chemicals such as acids, bases, alcohol, ketones, aromatics and more
40 x 50 cm
113 L
Absorbent Pad
100/Cs.
7.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.