missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Wide Neck Theraphthalate (PETG) Containers, Crystal-clear

Use the brilliant transparency of the Kautex™ Wide Neck Theraphthalate (PETG) Containers, Crystal-clear for a great product presentation.

Brand:  Kautex™ 2000074373

 View more versions of this product

Product Code. 10396301

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Series 310
  • Transparent PETG
  • Square with wide neck opening
  • Easy to fill and clean
  • Leakproof
  • Without closure
  • Ideal for product presentation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

200 mL
41 mm
Closure, Foam Insert
406
Black
Square
Wide Neck Container
Polyethylene
Transparent
Packaging, Dispensing, Sending Samples, Storage, Mailing
PET
406/Cs.
No
88 mm
Closure, Foam Insert
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions