Learn More
Bryson Packaging™ Polyethylene Bags
Product Code. 12350499
Click to view available options
Width (Metric):
100 mm
250 mm
300 mm
375 mm
Height (Metric):
150 mm
200 mm
250 mm
Quantity:
100 Pack
50 Pack
Unit Size:
Pack of 100
Pack of 50
Product Code. 12350499
Brand: Bryson Packaging™ BAJ300130L
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Transparent and excellent for shipping, storage and as sample bags.
  • Available in a wide range of sizes/capacities
  • Suitable for food use
Specifications
Description Bag
Material Polyethylene
Quantity 50 Pack
Width (Metric) 375 mm
Height (Metric) 250 mm