Learn More
Brady™ Polyester Thermal-Transfer Printable Labels
Polyester Thermal-Transfer Printable Labels are made of permanent polyester (B-423), which is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
POA - 662.00€
Specifications
Material Polyester
Print Technology Thermal Transfer
Product Type Thermal Transfer Printer Label
View More Specs
Products 33
Product Code Brand Shape Size (Metric) Finish Adhesive Type Price Quantity & Availability  
Product Code Brand Shape Size (Metric) Finish Adhesive Type Price Quantity & Availability  
13099262
View Documents
Brady™
030245
Rectangle 101.6W x 50.8mmH Gloss Acrylic, Permanent
POA
1 roll
Add to basket
10479883
View Documents Promotion Details
Brady™
217065
Round 9.53 mm dia. Matte Acrylic, Permanent
POA
1 roll
Add to basket
11504314
View Documents Promotion Details
Brady™
800536
Rectangle with Rounded Corners 40W x 15 mmH Matte Acrylic, Permanent
460.00€
1 roll
In Stock
to see stock.
Edge
Add to basket
13019302
View Documents Promotion Details
Brady™
030127
Rectangle 50.8W x 6.35 mmH Gloss Acrylic, Permanent
POA
1 roll
Add to basket
11896023
View Documents
Brady™
217087
Rectangle 19W x 38.1 mmH Matte Acrylic, Permanent
447.00€
1 roll
In Stock
to see stock.
Add to basket
13099302
View Documents Promotion Details
Brady™
030510
Rectangle 38.10W x 6.35mmH Gloss Acrylic, Permanent
POA
1 roll
Add to basket
11810855
View Documents
Brady™
030105
Rectangle 19 x 6.35 mm (W x H) Gloss Acrylic, Permanent
107.00€
1 roll
In Stock
to see stock.
Add to basket
11816043
View Documents Promotion Details
Brady™
217080
Rectangle 22.86W x 22.86 mmH Matte Acrylic, Permanent
307.00€
1 roll
In Stock
to see stock.
Add to basket
11583784
View Documents Promotion Details
Brady™
030221
Rectangle 25.4W x 4.75mmH Gloss Acrylic, Permanent
171.00€
1 roll
10209922
View Documents Promotion Details
Brady™
217071
Rectangle with Rounded Corners 25.4W x 9.53 mmH Matte Acrylic, Permanent
236.00€
1 roll
In Stock
to see stock.
Add to basket
11836013
View Documents
Brady™
217077
Rectangle 5.08W x 22.86 mmH Matte Acrylic, Permanent
331.00€
1 roll
In Stock
to see stock.
Add to basket
13009312
View Documents
Brady™
031337
Rectangle 25.4W x 12.7mmH Gloss Rubber, Permanent
277.00€
1 pack
In Stock
to see stock.
Add to basket
12834723
View Documents Promotion Details
Brady™
236719
Rectangle 50.8W x 25.4 mmH Satin Acrylic, Permanent
125.00€
1 roll
In Stock
to see stock.
Add to basket
10020942
View Documents Promotion Details
Brady™
217093
Rectangle 15.24 x 48.26 mm (W x H) Matte Acrylic, Permanent
518.00€
1 roll
In Stock
to see stock.
Add to basket
11538754
View Documents Promotion Details
Brady™
236713
Rectangle 25.4W x 76.2mmH Satin Acrylic, Permanent
POA
1 roll
Add to basket
11836023
View Documents
Brady™
217089
Rectangle 34.93W x 45.72 mmH Matte Acrylic, Permanent
POA
1 roll
Add to basket
11523724
View Documents
Brady™
030134
Rectangle 76.20W x 25.4mmH Gloss Acrylic, Permanent
320.00€
1 roll
In Stock
to see stock.
Add to basket
10011932
View Documents
Brady™
217070
Rectangle with Rounded Corners 15.24 mm dia. Matte Acrylic, Permanent
POA
1 roll
Add to basket
11814714
View Documents Promotion Details
Brady™
030137
Rectangle 76.20W x 50.8 mmH Gloss Acrylic, Permanent
214.00€
1 roll
In Stock
to see stock.
Add to basket
11503734
View Documents
Brady™
030142
Rectangle 31.75W x 19 mmH Gloss Acrylic, Permanent
POA
1 roll
Add to basket
11836043
View Documents Promotion Details
Brady™
236715
Rectangle 38.1W x 19 mmH Satin Acrylic, Permanent
141.00€
1 roll
In Stock
to see stock.
Add to basket
10031932
View Documents Promotion Details
Brady™
217088
Rectangle 15.24W x 38.1 mmH Matte Acrylic, Permanent
401.00€
1 roll
In Stock
to see stock.
Add to basket
10630134
View Documents Promotion Details
Brady™
030131
Rectangle 50.8 x 25.4 mm (W x H) Gloss Acrylic, Permanent
224.00€
1 roll
In Stock
to see stock.
Add to basket
10315992
View Documents Promotion Details
Brady™
217091
Rectangle 48.26W x 6.35 mmH Matte Acrylic, Permanent
283.00€
1 roll
In Stock
to see stock.
Add to basket
11885245
View Documents
Brady™
030104
Rectangle 25.4 x 12.70 mm (W x H) Gloss Acrylic, Permanent
231.00€
1 roll
In Stock
to see stock.
Add to basket
10600784
View Documents Promotion Details
Brady™
217072
Rectangle 41.28 x 15.24 mm (W xH) Matte Acrylic, Permanent
351.00€
1 roll
In Stock
to see stock.
Add to basket
11582715
View Documents
Brady™
014943
Rectangle 76.2W x 25.4mmH Gloss Acrylic, Permanent
384.00€
1 roll
10679343
View Documents
Brady™
217097
Rectangle 88.9W x 38.1 mmH Matte Acrylic, Permanent
POA
1 roll
In Stock
to see stock.
Add to basket
10561515
View Documents
Brady™
217092
Rectangle 18.26W x 25.46 mmH Matte Acrylic, Permanent
662.00€
1 roll
In Stock
to see stock.
Add to basket
11886013
View Documents Promotion Details
Brady™
217076
Rectangle 15.24 x 66.68 mm (W x H) Matte Acrylic, Permanent
378.00€
1 roll
In Stock
to see stock.
Add to basket
12882699
View Documents
Brady™
217079
Rectangle 22.86W x 19 mmH Matte Acrylic, Permanent
283.00€
1 roll
In Stock
to see stock.
Add to basket
11826033
View Documents
Brady™
217095
Rectangle 57.15W x 19 mmH Matte Acrylic, Permanent
615.00€
1 roll
In Stock
to see stock.
Add to basket
11826043
View Documents
Brady™
236701
Rectangle 19W x 6.35 mmH Satin Acrylic, Permanent
77.05€
1 roll
In Stock
to see stock.
Add to basket
Description
Description
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
  • Service temperature of -94 to 230° F (-70 to 110° C)
Specifications
Specifications
Polyester
Thermal Transfer
Thermal Transfer Printer Label