Learn More
3M™ Polycarbonate Visitor Glasses
Click to view available options
UV Protection
Description
- Lightweight polycarbonate lenses
- Moulded sideshields and browguards
- Can be worn on their own or over current style prescription spectacles.
- Integrated side-shield for maximum protection
- Optical class 1 for maximum clarity
- Lens coatings provide scratch resistance (Visitor Type 3)
Specifications
Specifications
Certifications/Compliance | EN166-1F |
Product Type | Protective eyewear |
For Use With (Application) | Protection against low energy impact, UV protection |
Lens Material | Polycarbonate |
UV Protection | Yes |