Learn More
Fisherbrand™ Polycarbonate Membranes
Fisherbrand™ Polycarbonate Membranes smooth flat surface and hexibit very low extractables levels.
Brand:  Fisherbrand™ 0482A00023
Product Code. 11785315
  • 335.00€ / Pack of 100
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Track-etched PC membranes manufactured from high-quality polycarbonate film
Smooth flat surface and hexibit very low extractables levelsFeatures high flow rates and effective chemical and thermal resistance due to sharply defined pore sizesSuitable for detection of particles in many corrosive fluids
Specifications
Specifications
White
0.2 μm
10mL/min./cm2
3L/min./cm2
0.25 mm
Yes
Polycarbonate
Non-sterile
Fluids clarification, environnemental analysis, cytology, parasitology, bioassays, epifluorescence microscopy, analysis of biological cells and particles
100 Pack
Plain Disc
47 mm