Learn More
Masterflex™ Polycarbonate Male Luer Adapter
Product Code. p-8004464
Click to view available options
For Use With:
Fits Male Luer Lock x 1/16 in. I.D. Low Profile Barb
Fits Male luer with lock ring x 3/16 in. hose barb
Unit Size:
Pack of 25
Product Code. 15354414
Brand: Masterflex™ 4550407
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Material Polycarbonate
For Use With Fits Male Luer Lock x 1/16 in. I.D. Low Profile Barb
Packaging Quantity 25 Pack
Product Type Luer Adapter