accessibility menu, dialog, popup
Test
chevron-down
Select Language
English
French
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Specialty Lab Equipment, Instruments, and Apparatuses
Laboratory Spatulas
VITLAB™ Polyamide Double Spoons
15292779
Print
VITLAB™ Polyamide Double Spoons
Length: 180mm
Brand:
VITLAB™ 80595
View more versions of this product
Product Code.
15292779
30.25€
/ Pack of 10
Quantity
Estimated Shipment: 17-01-2025
Log in
to see stock.
Explore more special offers
Specifications
Length (Metric)
180 mm