missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Euro Protection™ Polyamide Coated Gloves
Click to view available options
Size:
7
Specifications
Specifications
Certifications/Compliance | CE Mark |
Coated Area | Palm |
Coating Material | Nitrile Foam |
Color | Gray |
Material | Polyamide |
Product Type | Protective Gloves |
Size | 7 |
Palm Type | Gray Foam Nitrile |
Quantity | 1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction